Learn More
Thermco Products Inc NIST Traceable - Factory Certified Non-Mercury Glass Thermometers - Spirit Filled
Catalog No. NC1392437
Encompass_Preferred
Range: -20/110°C, Immersion: 76mm, and Length: 305mm.
Catalog No. NC1392437 Supplier Thermco Products Inc Supplier No. ACC6113SXYFC
Range: -20/110°C, Immersion: 76mm, and Length: 305mm.