Learn More
Thermco Products Inc NIST TRACEABLE - Factory Certified Glass Thermometer, Spirit Filled
Catalog No. NC1392438
Encompass_Preferred
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-InGlass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case. Range: -20-110°C, Divisions: 1°C, Immersions: Total, Certified at: 0°C, 37°C, 56°C, Length: 305mm
Catalog No. NC1392438 Supplier Thermco Products Inc Supplier No. ACC611SSCFC
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-InGlass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case. Range: -20-110°C, Divisions: 1°C, Immersions: Total, Certified at: 0°C, 37°C, 56°C, Length: 305mm