Learn More
Thermco Products Inc Coliform Incubator Thermometer, 34.5/46C x 0.1C, Red Spirit, 76mm Imm, 240mm long
Catalog No. NC0738182
Encompass_Preferred
These spirit-filled glass thermometers meet EPA requirements. Coliform incubator thermometers are designed to use in incubators or baths during coliform tests. Use them to monitor equipment. Each includes an individual serial number and certificate of accuracy. The ACC713BLS can be certified and made traceable to N.I.S.T.
Catalog No. NC0738182 Supplier Thermco Products Inc Supplier No. ACC713BLS
These spirit-filled glass thermometers meet EPA requirements. Coliform incubator thermometers are designed to use in incubators or baths during coliform tests. Use them to monitor equipment. Each includes an individual serial number and certificate of accuracy. The ACC713BLS can be certified and made traceable to N.I.S.T.