Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Spiro[1-azabicyclo[2.2.2]octane-3,2'(3'H)-furo[2,3-b]pyridin]-5'-amine, (2'R)- | 220100-05-0 | 99.7% | 231.29 g·mol⁻¹ | C13H17N3O | 10 MG
SDP

Catalog No. 5000287394
Encompass_Preferred

α7 Nicotinic receptor agonist-1 is a small-molecule research compound that functions as an agonist of the α7 nicotinic acetylcholine receptor. Supplied as a solid for in vitro pharmacology and receptor assay studies, the compound is provided with analytical data and storage recommendations; it is intended for research use only and should be handled according to the applicable safety data sheet.

  • Acts as α7 nicotinic acetylcholine receptor agonist.
  • High purity (99.7%).
  • CAS number 220100-05-0.
  • Molecular formula C13H17N3O; molecular weight 231.29 g·mol⁻¹.
  • Light yellow to light brown solid appearance.
  • Storage: 4 °C, protect from light.
  • Provided with a certificate of analysis.

Catalog No. 50-002-87394 Supplier Medchemexpress LLC Supplier No. HY14832510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α7 Nicotinic receptor agonist-1 is a small-molecule research compound that functions as an agonist of the α7 nicotinic acetylcholine receptor. Supplied as a solid for in vitro pharmacology and receptor assay studies, the compound is provided with analytical data and storage recommendations; it is intended for research use only and should be handled according to the applicable safety data sheet.

  • Acts as α7 nicotinic acetylcholine receptor agonist.
  • High purity (99.7%).
  • CAS number 220100-05-0.
  • Molecular formula C13H17N3O; molecular weight 231.29 g·mol⁻¹.
  • Light yellow to light brown solid appearance.
  • Storage: 4 °C, protect from light.
  • Provided with a certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.