Learn More
Medchemexpress LLC Spiro[1H,3H-naphtho[1,8-cd]pyran-1,9'-[9H]xanthen]-3-one, 3',6'-dihydroxy- | 41307-63-5 | 99.9% | 25 MG
SDP
Catalog No. 5000231270
Encompass_Preferred
Resorcinolnaphthalein is a small-molecule research reagent (C24H14O5) used as an ACE2 enhancer in biochemical assays. It is supplied as a high-purity solid suitable for in vitro research.
  • Functions as an ACE2 enhancer with reported EC50 of 19.5 μM.
  • High purity as measured by LCMS (99.9%).
  • Provided as a light yellow to orange solid; available in solid quantities and as a 10 mM solution in DMSO.
  • Molecular weight 382.4 g/mol; formula C24H14O5.
  • Intended for research use only and not for human use.
Catalog No. 50-002-31270 Supplier Medchemexpress LLC Supplier No. HY12244525MG
Resorcinolnaphthalein is a small-molecule research reagent (C24H14O5) used as an ACE2 enhancer in biochemical assays. It is supplied as a high-purity solid suitable for in vitro research.
  • Functions as an ACE2 enhancer with reported EC50 of 19.5 μM.
  • High purity as measured by LCMS (99.9%).
  • Provided as a light yellow to orange solid; available in solid quantities and as a 10 mM solution in DMSO.
  • Molecular weight 382.4 g/mol; formula C24H14O5.
  • Intended for research use only and not for human use.