Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Spiro[9H-fluorene-9,9'-[9H]xanthene]-2,7-diamine

Catalog No. 9048991G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-489-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-489-91G Supplier Sigma Aldrich Supplier No. 9048991G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Spiro[9H-fluorene-9,9G-[9H]xanthene]-2,7-diamine (X59) is a hole transporting material (HTM), which has a spiro[fluorene-9,9G-xanthene] as a core component. It can be synthesized by Buchwald-Hartwig reaction. It shows a power conversion efficiency (PCE) of 19.8%.X59 is a new hole transporting material (HTM) with spiro[fluorene-9,9G-xanthene] as the core moiety. An impressive power conversion efficiency (PCE) of 19.8% was achieved by using X59 as HTM in perovskite solar cell, which can compete with the record PCE by using the state-of-the-art-HTM Spiro-OMeTAD. The X59-based devices show negligible hysteresis and reasonable stability in dark and dry conditions at room temperature for over five weeks.

Specifications

CAS 2095034-97-0
Molecular Formula C53H42N2O5
Linear Formula C53H42N2O5
Quantity 1 g
Synonym N,N,N,N-tetrakis(4-methoxyphenyl)spiro[fluorene-9,9 -xanthene]-2,7-diamine; X59
Molecular Weight (g/mol) 786.91
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.