Learn More
Cayman Chemical Spiro-MeOTAD, Purity: ≥95%, CAS: 207739-72-8, 10 mg
SDP
Catalog No. 502979627
Encompass_Preferred
A stable and efficient hole-transport material in organic light-emitting devices and in ssDSSCs; yields high ssDSSC efficiency due to its excellent pore-filling property into mesoporous titania films
Catalog No. 50-297-9627 Supplier Cayman Chemical Supplier No. 1731710 MG
A stable and efficient hole-transport material in organic light-emitting devices and in ssDSSCs; yields high ssDSSC efficiency due to its excellent pore-filling property into mesoporous titania films