Learn More
Newcomer Supply Spirochete, Artificial Control Slides, 98/set
SDP
Catalog No. NC0779289
Encompass
Newcomer Supply Spirochete, Artificial Control Slides are for the positive histochemical staining of spirochetes, the causative agent of a variety of diseases such as; syphilis, bejel, pinta, yaws and lyme. Brachyspira hyodysenteriae, purchased from American Type Culture Collection, is used to produce the positive control tissue. Spirochete, Artificial Control Slides contain sections of positive staining rat lung.
Catalog No. NC0779289 Supplier Newcomer Supply Supplier No. 4655B
Newcomer Supply Spirochete, Artificial Control Slides are for the positive histochemical staining of spirochetes, the causative agent of a variety of diseases such as; syphilis, bejel, pinta, yaws and lyme. Brachyspira hyodysenteriae, purchased from American Type Culture Collection, is used to produce the positive control tissue. Spirochete, Artificial Control Slides contain sections of positive staining rat lung.