Learn More
Cayman Chemical SpIrohexenolIde A 100ug
SDP
Catalog No. 502934478
Encompass_Preferred
A bacterial metabolite; inhibits growth in the NCI-60 cancer cell line panel (GI50s = 0.1-17 μM) with the RPMI8226 multiple myeloma, HOP-92 lung, and SW620 colon cancer cell lines being most sensitive (GI50s = 254, 191, and 565 nM, respectively); binds MIF (Kd = 35.9 μM); inhibits MIF cellular uptake and lysosomal localization in HCT116 cells
Catalog No. 50-293-4478 Supplier Cayman Chemical Supplier No. 25116100 UG
A bacterial metabolite; inhibits growth in the NCI-60 cancer cell line panel (GI50s = 0.1-17 μM) with the RPMI8226 multiple myeloma, HOP-92 lung, and SW620 colon cancer cell lines being most sensitive (GI50s = 254, 191, and 565 nM, respectively); binds MIF (Kd = 35.9 μM); inhibits MIF cellular uptake and lysosomal localization in HCT116 cells