Learn More
Medchemexpress LLC Spiroindoline-based tankyrase inhibitor | 2171388-28-4 | 98.1% | C27H35FN6O3 | 10MG
SDP
Catalog No. 5000258995
Encompass_Preferred
RK-582 is an orally active, spiroindoline-based selective tankyrase inhibitor with nanomolar potency and demonstrated antiproliferative activity in colorectal cancer models.
Catalog No. 50-002-58995 Supplier Medchemexpress LLC Supplier No. HY13784910MG
RK-582 is an orally active, spiroindoline-based selective tankyrase inhibitor with nanomolar potency and demonstrated antiproliferative activity in colorectal cancer models.