Learn More
Cayman Chemical Spirolaxine, Purity: ≥95%, CAS: 126382-01-2, 2.5 mg
SDP
Catalog No. 502987905
Encompass_Preferred
A phthalide antibacterial agent; strong anti-H. pylori effect (MIC = 0.2 µg/cm3); 1-40 µM inhibits LoVo and HL-60 cell proliferation
Catalog No. 50-298-7905 Supplier Cayman Chemical Supplier No. 220632.5 MG
A phthalide antibacterial agent; strong anti-H. pylori effect (MIC = 0.2 µg/cm3); 1-40 µM inhibits LoVo and HL-60 cell proliferation