Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Spirometry Calibration Syringe Cabinet
SDP

Catalog No. 22000711
Change view
Click to view available options
Material:
PETG
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
22-000-711 PETG
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-000-711 Supplier Poltex Supplier No. SPIROCALW
Add to Cart
Add to Cart
This item is not returnable. View return policy

Poltex Spirometry Calibration Syringe Cabinet fits Midmark or equivalent spirometry calibration syringe.

  • Holds spirometry calibration syringe.
  • Magnetic catch on door.
  • Removable keepers sit securely in slots to keep syringe in place.
  • Durable, cleanable PETG construction.
  • Fits Midmark or equivalent spirometry calibration syringe.
  • See image for dimensions.

Specifications

Autoclavable Not Autoclavable
Color Clear
Material PETG
Positions 1
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Midmark or Equivalent Sprirometry Calibration Syringe
Mounting Wall Mount
Length (Metric) 139 mm
Height (Metric) 596 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.