Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Spironolactone | 52-01-7 | MFCD00082250 | 98.6% | 416.57 g/mol | C24H32O4S | 10 MG
SDP

Catalog No. 5000299029
Encompass_Preferred

Spironolactone analytical reference standard intended for research and analytical applications, including qualitative and quantitative methods (HPLC, GC, MS). The product is supplied as a small-quantity solid with high stated purity to support accurate assay performance.

  • Analytical/reference standard suitable for HPLC, GC, and MS.
  • High stated purity (98.6%), supporting accurate quantitative results.
  • Molecular weight 416.57 g/mol for mass identification.
  • CAS number 52-01-7 for unambiguous compound identification.
  • Supplied in a 10 mg quantity for analytical and method development use.

Catalog No. 50-002-99029 Supplier Medchemexpress LLC Supplier No. HYB0561R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spironolactone analytical reference standard intended for research and analytical applications, including qualitative and quantitative methods (HPLC, GC, MS). The product is supplied as a small-quantity solid with high stated purity to support accurate assay performance.

  • Analytical/reference standard suitable for HPLC, GC, and MS.
  • High stated purity (98.6%), supporting accurate quantitative results.
  • Molecular weight 416.57 g/mol for mass identification.
  • CAS number 52-01-7 for unambiguous compound identification.
  • Supplied in a 10 mg quantity for analytical and method development use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.