Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spironolactone, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022889 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
150 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2889 150 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2889 Supplier MilliporeSigma Supelco Supplier No. BP306
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

TRUSTED_SUSTAINABILITY

Specifications

CAS 52-01-7
Melting Point 207°C to 208°C (literature)
Molecular Formula C24H32O4S
Linear Formula C24H32O4S
Quantity 150 mg
MDL Number MFCD00082250
Synonym 4-Pregnen-21-oic acid-17 alpha-ol-3-one-7 alpha-thiol gamma-lactone 7-acetate; 7 alpha-(Acetylthio)-17 alpha-hydroxy-3-oxopregn-4-ene-21-carboxylic acid gamma-lactone
Formula Weight 416.57
Grade BP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.