Learn More
Medchemexpress LLC Spironolactone-d3 | 52-01-7 | 99.82% | 1 MG
SDP
Catalog No. 5000407903
Encompass_Preferred
Spironolactone-d3 is the deuterium labeled Spironolactone, an orally active aldosterone mineralocorticoid receptor antagonist with an IC50 of 24 nM. It also acts as a potent antagonist of androgen receptor with an IC50 of 77 nM and promotes autophagy in podocytes.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07903 Supplier Medchemexpress LLC Supplier No. HYB0561S11MG
Spironolactone-d3 is the deuterium labeled Spironolactone, an orally active aldosterone mineralocorticoid receptor antagonist with an IC50 of 24 nM. It also acts as a potent antagonist of androgen receptor with an IC50 of 77 nM and promotes autophagy in podocytes.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS