Learn More
Medchemexpress LLC Spironolactone-d7 | 100.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0561S1MG
Encompass_Preferred
Spironolactone-d7 is the deuterium-labeled version of Spironolactone, an orally active aldosterone mineralocorticoid receptor antagonist with an IC50 of 24 nM. It also acts as a potent antagonist of the androgen receptor with an IC50 of 77 nM and promotes autophagy in podocytes.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Intended for research use only.
Catalog No. 50-004-07905