Learn More
Cayman Chemical SpIrotetramat 100mg
SDP
Catalog No. 502936655
Encompass_Preferred
An insecticide; increases the activity of GPX and SOD and decreases levels of MDA in Chinese toad tadpoles at 0.06 mg/L; induces toxicity in Chinese toad tadpoles (LC50 = 6.98 mg/L)
Catalog No. 50-293-6655 Supplier Cayman Chemical Supplier No. 25634100 MG
An insecticide; increases the activity of GPX and SOD and decreases levels of MDA in Chinese toad tadpoles at 0.06 mg/L; induces toxicity in Chinese toad tadpoles (LC50 = 6.98 mg/L)