Learn More
Cayman Chemical SpIroxamIn-d4 1mg
SDP
Catalog No. 502918351
Encompass_Preferred
An internal standard for the quantification of spiroxamine by GC- or LC-MS
Catalog No. 50-291-8351 Supplier Cayman Chemical Supplier No. 396041 MG
Add to Cart
Add to Cart
An internal standard for the quantification of spiroxamine by GC- or LC-MS