Learn More
Medchemexpress LLC Spiroxamine | 118134-30-8 | 99.9% | 50 MG
SDP
Catalog No. 5000230899
Encompass_Preferred
Spiroxamine analytical standard (CAS 118134-30-8) is supplied as a high-purity liquid reference material for analytical and residue testing. It is intended for method development, calibration, and quantitative analyses using HPLC, GC, and MS.
  • High purity of 99.9% (COA available).
  • Suitable as an analytical reference for HPLC, GC, and MS.
  • Molecular formula C18H35NO2; molecular weight 297.48 g/mol.
  • Available in small pack sizes suitable for analytical workflows.
  • SDS and certificate of analysis available for quality verification.
Catalog No. 50-002-30899 Supplier Medchemexpress LLC Supplier No. HY121884R50MG
Spiroxamine analytical standard (CAS 118134-30-8) is supplied as a high-purity liquid reference material for analytical and residue testing. It is intended for method development, calibration, and quantitative analyses using HPLC, GC, and MS.
  • High purity of 99.9% (COA available).
  • Suitable as an analytical reference for HPLC, GC, and MS.
  • Molecular formula C18H35NO2; molecular weight 297.48 g/mol.
  • Available in small pack sizes suitable for analytical workflows.
  • SDS and certificate of analysis available for quality verification.