Learn More
Medchemexpress LLC Spisulosine-d3 (ES-285-d3) | 1246298-31-6 | 99.0% | 288.53 | 1 MG
SDP
Catalog No. 5000351897
Encompass_Preferred
Spisulosine-d3 (ES-285-d3) is a deuterium-labeled version of Spisulosine, an antiproliferative compound derived from marine sources. It inhibits the growth of prostate PC-3 and LNCaP cells by causing intracellular ceramide accumulation, activating PKCζ, and inducing apoptosis. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-51897 Supplier Medchemexpress LLC Supplier No. HY13626S1MG
Spisulosine-d3 (ES-285-d3) is a deuterium-labeled version of Spisulosine, an antiproliferative compound derived from marine sources. It inhibits the growth of prostate PC-3 and LNCaP cells by causing intracellular ceramide accumulation, activating PKCζ, and inducing apoptosis. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Can affect pharmacokinetic and metabolic profiles of drugs