Learn More
Biolegend Spark NIR™ 685 anti-mouse CD19
Supplier:  Biolegend 115568
Encompass_Preferred
Spark NIR™ 685 anti-mouse CD19 [6D5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-0201