Learn More
Biolegend Spark YG™ 570 anti-mouse/human PNAd
Supplier:  Biolegend 120809
Encompass_Preferred
Spark YG™ 570 anti-mouse/human PNAd [MECA-79]; Isotype: Rat IgM, κ; Reactivity: Mouse, Human; Apps: IHC-F, IHC-P; Size: 25 ug
Catalog No. 50-207-0430