Learn More
Biolegend Spark YG™ 570 anti-Tyrosine Hydroxylase
Supplier:  Biolegend 818009
Encompass_Preferred
Spark YG™ 570 anti-Tyrosine Hydroxylase [2/40/15]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P; Size: 25 ug
Catalog No. 50-207-4111