Learn More
PRO LAB SUPPLY CORP SPL SEPTUM TUBE 50 ML CS/25
SDP
Catalog No. NC2749741
Encompass_Preferred
SPL Septum Tube 50 ml, Septum (Silicone/PTFE) RCF Rating 13,000 x g, Sterile Case of 25  
Catalog No. NC2749741 Supplier PRO LAB SUPPLY CORP Supplier No. 50750
SPL Septum Tube 50 ml, Septum (Silicone/PTFE) RCF Rating 13,000 x g, Sterile Case of 25  