Learn More
World Precision Instrument SPL Syringe Pump - Programmable Infuse/Withdrawl
SDP
Catalog No. 502538568
Encompass
The large touch screen color display lets you see all of the pump’s operating parameters to ensure proper operation during the experiments. Syringe size and flow rate are easily displayed, as well as the volume delivered and elapsed time. Set up is easy using the icon-driven software. An LED on the front panel makes it easy to see if the pump is running. Advanced micro stepping techniques are employed to further reduce the step angle to eliminate flow pulsation. Accuracy is ±0.5%. A wide dynamic flow range from picoliters per minute to millimeters per minute can be programmed into the pump. These versatile pumps can be connected through an RS485 interface. Add the new Adagio software to maximize the use of the pump’s functions and features. Adagio allows you to configure the pump through the software, as well as operate one or multiple pumps. LabVIEW drivers are available on the National Instruments website.
Catalog No. 50-253-8568 Supplier World Precision Instrument Supplier No. SPLG212
Add to Cart
Add to Cart
Request a Quote
The large touch screen color display lets you see all of the pump’s operating parameters to ensure proper operation during the experiments. Syringe size and flow rate are easily displayed, as well as the volume delivered and elapsed time. Set up is easy using the icon-driven software. An LED on the front panel makes it easy to see if the pump is running. Advanced micro stepping techniques are employed to further reduce the step angle to eliminate flow pulsation. Accuracy is ±0.5%. A wide dynamic flow range from picoliters per minute to millimeters per minute can be programmed into the pump. These versatile pumps can be connected through an RS485 interface. Add the new Adagio software to maximize the use of the pump’s functions and features. Adagio allows you to configure the pump through the software, as well as operate one or multiple pumps. LabVIEW drivers are available on the National Instruments website.