Learn More
Enzo Life Sciences sPLA2 activity kit (1x96wells)
SDP
Catalog No. 50658602
Encompass_Preferred
The sPLA2 activity kit is a higher throughput assay for the quantification of up to 42 samples in duplicate per 96 well kit. This kit provides safe, sensitive, and reliable results in under an hour. Alternative Name: Secretory phospholipase A2. Sensitivity: 2.34 Units/ml (range 5 - 80 Units/mL). Applications: Colorimetric detection, Activity assay. Contents: Microtiter plate, Standard, Reaction buffer concentrate, Substrate, Stop reagent, Color reagent
Catalog No. 50-658-602 Supplier Enzo Life Sciences Supplier No. ADI907002
The sPLA2 activity kit is a higher throughput assay for the quantification of up to 42 samples in duplicate per 96 well kit. This kit provides safe, sensitive, and reliable results in under an hour. Alternative Name: Secretory phospholipase A2. Sensitivity: 2.34 Units/ml (range 5 - 80 Units/mL). Applications: Colorimetric detection, Activity assay. Contents: Microtiter plate, Standard, Reaction buffer concentrate, Substrate, Stop reagent, Color reagent