Learn More
Cayman Chemical sPLA2 asy Buffer10X 1ea
SDP
Catalog No. 502896266
Encompass_Preferred
An internal standard for the quantification of omeprazole sulfide by GC- or LC-MS
Catalog No. 50-289-6266 Supplier Cayman Chemical Supplier No. 7650101 EA
An internal standard for the quantification of omeprazole sulfide by GC- or LC-MS