Learn More
Cayman Chemical sPLA2 asy KIt 96Wel
SDP
Catalog No. 502894816
Encompass_Preferred
Cayman’s sPLA2 Assay provides an accurate and convenient method for measurement of sPLA2 activity. This assay uses the 1,2-dithio analog of diheptanoyl phosphatidylcholine which serves as a substrate for most PLA2s with the exception of cPLA2. Upon hydrolysis of the thioester bond at the sn-2 position by PLA2, free thiols are detected using DTNB.
Catalog No. 50-289-4816 Supplier Cayman Chemical Supplier No. 76500196 WELL
Add to Cart
Add to Cart
Cayman’s sPLA2 Assay provides an accurate and convenient method for measurement of sPLA2 activity. This assay uses the 1,2-dithio analog of diheptanoyl phosphatidylcholine which serves as a substrate for most PLA2s with the exception of cPLA2. Upon hydrolysis of the thioester bond at the sn-2 position by PLA2, free thiols are detected using DTNB.