Learn More
Cayman Chemical sPLA2 InhIbItr 10mg
SDP
Catalog No. 502966758
Encompass_Preferred
An orally active inhibitor of sPLA2-IIA; protects against intestinal reperfusion injury in rats when given at 10 mg/kg orally; attenuates NF-κB signaling in lung cancer cells and protects against diet-induced metabolic syndrome in rats
Catalog No. 50-296-6758 Supplier Cayman Chemical Supplier No. 1727710 MG
Add to Cart
Add to Cart
An orally active inhibitor of sPLA2-IIA; protects against intestinal reperfusion injury in rats when given at 10 mg/kg orally; attenuates NF-κB signaling in lung cancer cells and protects against diet-induced metabolic syndrome in rats