Learn More
Cayman Chemical sPLA2 Inhibitor, Purity: ≥95%, CAS: 393569-31-8, 1 mg
SDP
Catalog No. 502967460
Encompass_Preferred
An orally active inhibitor of sPLA2-IIA; protects against intestinal reperfusion injury in rats when given at 10 mg/kg orally; attenuates NF-κB signaling in lung cancer cells and protects against diet-induced metabolic syndrome in rats
Catalog No. 50-296-7460 Supplier Cayman Chemical Supplier No. 172771 MG
An orally active inhibitor of sPLA2-IIA; protects against intestinal reperfusion injury in rats when given at 10 mg/kg orally; attenuates NF-κB signaling in lung cancer cells and protects against diet-induced metabolic syndrome in rats