Learn More
Cayman Chemical sPLA2 InhIbItr 500ug
SDP
Catalog No. 502967553
Encompass_Preferred
An orally active inhibitor of sPLA2-IIA; protects against intestinal reperfusion injury in rats when given at 10 mg/kg orally; attenuates NF-κB signaling in lung cancer cells and protects against diet-induced metabolic syndrome in rats
Catalog No. 50-296-7553 Supplier Cayman Chemical Supplier No. 17277500 UG
An orally active inhibitor of sPLA2-IIA; protects against intestinal reperfusion injury in rats when given at 10 mg/kg orally; attenuates NF-κB signaling in lung cancer cells and protects against diet-induced metabolic syndrome in rats