Learn More
Cayman Chemical sPLA2mouse TYP V Blockng 200ug
SDP
Catalog No. 502911908
Encompass_Preferred
A PI4KIIIa inhibitor (IC50 3 16 nM) selective for PI4KIIIa over PI4KIIIB PI3Ka PI3KB and PI3K (IC50s 50 nM) inhibits PI3KY (IC50 15 8 nM) decreases the levels of PtdIns-(4)-P1 in HEK293 cells expressing the AT1 receptor (IC50 3 nM) inhibits HCV genotype 1a and 1b replication
Catalog No. 50-291-1908 Supplier Cayman Chemical Supplier No. 360512200 UG
A PI4KIIIa inhibitor (IC50 3 16 nM) selective for PI4KIIIa over PI4KIIIB PI3Ka PI3KB and PI3K (IC50s 50 nM) inhibits PI3KY (IC50 15 8 nM) decreases the levels of PtdIns-(4)-P1 in HEK293 cells expressing the AT1 receptor (IC50 3 nM) inhibits HCV genotype 1a and 1b replication