Learn More
Chemglass Life Sciences Top Outer:14/20, Lower Inner:14/20
SDP
Catalog No. 50872856
Encompass_Preferred
Adapter, splash guard, with fritted disc. Top Outer: 14/20, Lower Inner: 14/20. Similar to CG-1041, but with the addition of an Extra Coarse fritted disc between the lower inner and top outer joint.
Catalog No. 50-872-856 Supplier Chemglass Life Sciences Supplier No. CG1041F01
Adapter, splash guard, with fritted disc. Top Outer: 14/20, Lower Inner: 14/20. Similar to CG-1041, but with the addition of an Extra Coarse fritted disc between the lower inner and top outer joint.