Learn More
Chemglass Life Sciences Top Outer: 24/40 Lower Inner: 24/40
SDP
Catalog No. 50872858
Encompass_Preferred
Adapter, splash guard, with fritted disc. Top Outer: 24/40 Lower Inner: 24/40. Similar to CG-1041, but with the addition of an Extra Coarse fritted disc between the lower inner and top outer joint.
Catalog No. 50-872-858 Supplier Chemglass Life Sciences Supplier No. CG1041F03
Adapter, splash guard, with fritted disc. Top Outer: 24/40 Lower Inner: 24/40. Similar to CG-1041, but with the addition of an Extra Coarse fritted disc between the lower inner and top outer joint.