Learn More
Chemglass Life Sciences Top Outer: 29/26 Lower Inner: 29/26
SDP
Catalog No. 50872859
Encompass_Preferred
Adapter, splash guard, with fritted disc. Top Outer: 29/26 Lower Inner: 29/26. Similar to CG-1041, but with the addition of an Extra Coarse fritted disc between the lower inner and top outer joint.
Catalog No. 50-872-859 Supplier Chemglass Life Sciences Supplier No. CG1041F04
Adapter, splash guard, with fritted disc. Top Outer: 29/26 Lower Inner: 29/26. Similar to CG-1041, but with the addition of an Extra Coarse fritted disc between the lower inner and top outer joint.