Learn More
IMEB, Inc Small Pathology Bone Band Saw Optional Splash Guard Cover
SDP
Catalog No. NC0376688
Encompass_Preferred
Pictured is a Splash Guard for IMEB Bone Band Saw (IMEB Bone Band Saw sold separately). IMEB Bone Band Saw is an efficient and economical way of cutting bone in laboratory. The bone band saw is only 19" high and has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust and particles.
Catalog No. NC0376688 Supplier IMEB, Inc Supplier No. SG666
Pictured is a Splash Guard for IMEB Bone Band Saw (IMEB Bone Band Saw sold separately). IMEB Bone Band Saw is an efficient and economical way of cutting bone in laboratory. The bone band saw is only 19" high and has a foot print of 17" x 13" of counter space. This saw has a separate water-cooled blade that eliminates dust and particles.