Learn More
Medicus Health Splash Shield BioHazard Benchtop, Large
SDP
Supplier:  Medicus Health 2484M2
Encompass
Large Biohazard Benchtop Safety Shield allows space for to tube racks at the same time increasing your production as well as your splash protection∙ Biohazard Safety Splash Shield increases splash protection by keeping hazardous material contained while you work∙ Dimensions: 15""""W x 12""""D x 20""""H ∙ Durable, scratch-resistant plexiglass shield protects you from splashes, aerosol mist, and flying debris∙ Economic alternative provides great splash protection∙ Helps meet compliance with OSHA bloodborne pathogen regulations (29 CFR Part 191.1030)∙ Our Lab Shields are made from durable PETG plastic that's 20 times stronger than acrylic substitute shields which crack, splinter, craze and yellow easily - accept no substitutes∙ Clear shield provides easy viewing and can be cleaned with any standard laboratory disinfectant∙ A nice addition to a fume hood∙ Skid resistant feet keep shield in place and make it easy to position
Catalog No. NC2372229