Learn More
STEMCELL Technologies Spleen Dissociation Medium, Size: 40 mL
SDP
Catalog No. NC0676113
Encompass
This product, containing enzymes, is designed to maximize the viability and recovery of dendritic cells from mouse spleen when used with EasySep™ cell separation technology. A single 4 mL tube is sufficient for processing up to two mouse spleens.
Catalog No. NC0676113 Supplier STEMCELL Technologies Supplier No. 07915
This product, containing enzymes, is designed to maximize the viability and recovery of dendritic cells from mouse spleen when used with EasySep™ cell separation technology. A single 4 mL tube is sufficient for processing up to two mouse spleens.