Learn More
Meso Scale Discovery SPLEX HUM CTLA4 Kit 25 PL
SDP
Catalog No. 5000078919
Encompass
The Meso Scale Discovery CTLA-4 Discovery Kit is a highly sensitive immunoassay that quantitatively measures soluble CTLA-4 in serum plasma and cell culture supernatants The assay uses MSD s electrochemiluminescent detection technology and provides a lower limit of detection of 0 05 ng/mL The kit includes a 96-well plate coated with capture antibody a detection antibody conjugated to an electrochemiluminescent label and calibrators and controls to generate a standard curve The assay takes approximately 3 5 hours to complete and requires a MSD instrument such as SECTOR Imager QuickPlex or Discovery Workbench to read the plate The kit can test up to 80 samples in duplicate
Catalog No. 50-000-78919 Supplier Meso Scale Discovery Supplier No. K151AGLS4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery CTLA-4 Discovery Kit is a highly sensitive immunoassay that quantitatively measures soluble CTLA-4 in serum plasma and cell culture supernatants The assay uses MSD s electrochemiluminescent detection technology and provides a lower limit of detection of 0 05 ng/mL The kit includes a 96-well plate coated with capture antibody a detection antibody conjugated to an electrochemiluminescent label and calibrators and controls to generate a standard curve The assay takes approximately 3 5 hours to complete and requires a MSD instrument such as SECTOR Imager QuickPlex or Discovery Workbench to read the plate The kit can test up to 80 samples in duplicate