Learn More
Meso Scale Discovery SPLEX HUM CTLA4 KitQP 25 PL
SDP
Catalog No. 5000079486
Encompass
The S-PLEX Human CTLA-4 Kit by Meso Scale Diagnostics is a multiplex immunoassay kit that enables the simultaneous measurement of CTLA-4 in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and controls The electrochemiluminescent assay has a typical sensitivity of 0 3 pg/mL and an assay range from 0 3-2500 pg/mL The kit is for research use only and not for diagnostic procedures It allows high-throughput quantitative analysis of CTLA-4 with low sample volume requirements
Catalog No. 50-000-79486 Supplier Meso Scale Discovery Supplier No. K151AGLS24
Add to Cart
Add to Cart
Request a Quote
The S-PLEX Human CTLA-4 Kit by Meso Scale Diagnostics is a multiplex immunoassay kit that enables the simultaneous measurement of CTLA-4 in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and controls The electrochemiluminescent assay has a typical sensitivity of 0 3 pg/mL and an assay range from 0 3-2500 pg/mL The kit is for research use only and not for diagnostic procedures It allows high-throughput quantitative analysis of CTLA-4 with low sample volume requirements