Learn More
Meso Scale Discovery SPLEX HUM GCSF KitQP 25 PL
SDP
Catalog No. 5000079272
Encompass
The Meso Scale Discovery S-PLEX Human G-CSF Kit is a multiplex immunoassay that quantitatively measures Granulocyte Colony Stimulating Factor (G-CSF) in human serum plasma and cell culture supernatant samples The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less of sample and can perform up to 80 tests across two plates The lower limit of detection is 1 5 pg/mL
Catalog No. 50-000-79272 Supplier Meso Scale Discovery Supplier No. K151K3S24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human G-CSF Kit is a multiplex immunoassay that quantitatively measures Granulocyte Colony Stimulating Factor (G-CSF) in human serum plasma and cell culture supernatant samples The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less of sample and can perform up to 80 tests across two plates The lower limit of detection is 1 5 pg/mL