Learn More
Meso Scale Discovery SPLEX HUM GMCSF Kit 25 PL
SDP
Catalog No. 5000081983
Encompass
The S-PLEX Human GM-CSF Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of granulocyte-macrophage colony-stimulating factor (GM-CSF) in human serum plasma and cell culture supernatants The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires only a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and a 10X read buffer The assay can detect GM-CSF concentrations from 0 04 - 1500 pg/mL and has high specificity with minimal cross-reactivity to related analytes
Catalog No. 50-000-81983 Supplier Meso Scale Discovery Supplier No. K151F3S4
Add to Cart
Add to Cart
Request a Quote
The S-PLEX Human GM-CSF Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of granulocyte-macrophage colony-stimulating factor (GM-CSF) in human serum plasma and cell culture supernatants The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a broad dynamic range and requires only a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and a 10X read buffer The assay can detect GM-CSF concentrations from 0 04 - 1500 pg/mL and has high specificity with minimal cross-reactivity to related analytes