Learn More
Meso Scale Discovery SPLEX HUM GMCSF KitQP 25 PL
SDP
Catalog No. 5000082013
Encompass
The Meso Scale Discovery S-Plex Human GM-CSF Kit is a multiplex immunoassay for measuring GM-CSF in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection for high sensitivity broad dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies calibrators and necessary reagents It quantifies GM-CSF from 0 06 to 1500 pg/mL with high specificity and minimal cross-reactivity enabling efficient and accurate measurement for research
Catalog No. 50-000-82013 Supplier Meso Scale Discovery Supplier No. K151F3S24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-Plex Human GM-CSF Kit is a multiplex immunoassay for measuring GM-CSF in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection for high sensitivity broad dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies calibrators and necessary reagents It quantifies GM-CSF from 0 06 to 1500 pg/mL with high specificity and minimal cross-reactivity enabling efficient and accurate measurement for research