Learn More
Meso Scale Discovery SPLEX HUM IFN 2a Kit 25 PL
SDP
Catalog No. 5000078655
Encompass
The S-PLEX Human IFN alpha2a Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interferon alpha2a (IFN- 2a) in serum plasma cell culture supernatants and other biological matrices The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent calibrators and controls The lower limit of detection is 0 08 pg/mL and the upper limit of quantitation is 1500 pg/mL The assay takes approximately 3 hours to complete
Catalog No. 50-000-78655 Supplier Meso Scale Discovery Supplier No. K151P3S4
Add to Cart
Add to Cart
Request a Quote
The S-PLEX Human IFN alpha2a Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interferon alpha2a (IFN- 2a) in serum plasma cell culture supernatants and other biological matrices The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent calibrators and controls The lower limit of detection is 0 08 pg/mL and the upper limit of quantitation is 1500 pg/mL The assay takes approximately 3 hours to complete