Learn More
Meso Scale Discovery SPLEX HUM IFNB KitQP 25 PL
SDP
Catalog No. 5000079025
Encompass
The S-PLEX Human IFN beta Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring human interferon beta (IFN- ) in various biological matrices It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive quantitation of IFN- with detection limits from 0 04 to 400 pg/mL The kit includes a 96-well plate detection antibody calibrators and diluent The assay runs in 3 5 hours and requires 25 L of sample
Catalog No. 50-000-79025 Supplier Meso Scale Discovery Supplier No. K151ADRS24
Add to Cart
Add to Cart
Request a Quote
The S-PLEX Human IFN beta Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring human interferon beta (IFN- ) in various biological matrices It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive quantitation of IFN- with detection limits from 0 04 to 400 pg/mL The kit includes a 96-well plate detection antibody calibrators and diluent The assay runs in 3 5 hours and requires 25 L of sample