Learn More
Meso Scale Discovery SPLEX HUM IL10 KitQP 25 PL
SDP
Catalog No. 5000078779
Encompass
The S-PLEX Human IL-10 Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interleukin 10 (IL-10) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and controls The electrochemiluminescent assay has a sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL The total assay time is 3 5 hours This kit can be used to measure IL-10 levels in human samples for immunology and inflammation research
Catalog No. 50-000-78779 Supplier Meso Scale Discovery Supplier No. K151Y2S24
Add to Cart
Add to Cart
Request a Quote
The S-PLEX Human IL-10 Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human interleukin 10 (IL-10) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and controls The electrochemiluminescent assay has a sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL The total assay time is 3 5 hours This kit can be used to measure IL-10 levels in human samples for immunology and inflammation research