Learn More
Meso Scale Discovery SPLEX HUM IL12p70 KitQP
SDP
Catalog No. 5000078775
Encompass
The Meso Scale Discovery S-PLEX Human IL-12p70 Kit is an immunoassay that enables the simultaneous detection of IL-12p70 in human serum plasma and culture supernatants This multiplex assay kit utilizes MSD s electrochemiluminescent detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The kit includes a plate coated with capture antibodies detection antibodies and calibrators ranging from 0 61 pg/mL to 10 000 pg/mL The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL This product allows for the accurate sensitive and rapid quantification of IL-12p70 from human samples
Catalog No. 50-000-78775 Supplier Meso Scale Discovery Supplier No. K151G3S24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human IL-12p70 Kit is an immunoassay that enables the simultaneous detection of IL-12p70 in human serum plasma and culture supernatants This multiplex assay kit utilizes MSD s electrochemiluminescent detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The kit includes a plate coated with capture antibodies detection antibodies and calibrators ranging from 0 61 pg/mL to 10 000 pg/mL The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL This product allows for the accurate sensitive and rapid quantification of IL-12p70 from human samples