Learn More
Meso Scale Discovery SPLEX HUM IL13 KitQP 25 PL
SDP
Catalog No. 5000078930
Encompass
The Meso Scale Discovery S-PLEX Human IL-13 Kit is a multiplex immunoassay for measuring IL-13 in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies buffers diluent and calibrators The assay takes 3 5 hours and requires a MESO QuickPlex SQ 120 instrument It quantifies IL-13 important in immune responses
Catalog No. 50-000-78930 Supplier Meso Scale Discovery Supplier No. K151Y9S24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human IL-13 Kit is a multiplex immunoassay for measuring IL-13 in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies buffers diluent and calibrators The assay takes 3 5 hours and requires a MESO QuickPlex SQ 120 instrument It quantifies IL-13 important in immune responses