Learn More
Meso Scale Discovery SPLEX HUM IL17A Kit 25 PL
SDP
Catalog No. 5000082055
Encompass
The Meso Scale Discovery S-PLEX Human IL-17A Kit is a multiplex immunoassay that measures IL-17A levels in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer diluent and calibrators to generate a standard curve The assay takes 3 5 hours to complete and has a detection range of 0 06-2500 pg/mL
Catalog No. 50-000-82055 Supplier Meso Scale Discovery Supplier No. K151C3S4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human IL-17A Kit is a multiplex immunoassay that measures IL-17A levels in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium read buffer wash buffer diluent and calibrators to generate a standard curve The assay takes 3 5 hours to complete and has a detection range of 0 06-2500 pg/mL