Learn More
Meso Scale Discovery SPLEX HUM IL1B KitQP 25 PL
SDP
Catalog No. 5000079503
Encompass
The S-PLEX Human IL-1 Kit from Meso Scale Discovery is a highly sensitive immunoassay that enables the simultaneous detection of IL-1 in human serum plasma and cell culture supernatants This multiplex assay utilizes MSD s proprietary SECTOR Imager and SULFO-TAG labels for electrochemiluminescent detection The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium calibrators and reagents required to perform up to 25 tests in duplicate The assay has a sensitivity of 0 04 pg/mL and a broad dynamic range from 0 06 - 1500 pg/mL This kit allows for the rapid and accurate quantification of IL-1 levels in various human sample types
Catalog No. 50-000-79503 Supplier Meso Scale Discovery Supplier No. K151ADSS24
Add to Cart
Add to Cart
Request a Quote
The S-PLEX Human IL-1 Kit from Meso Scale Discovery is a highly sensitive immunoassay that enables the simultaneous detection of IL-1 in human serum plasma and cell culture supernatants This multiplex assay utilizes MSD s proprietary SECTOR Imager and SULFO-TAG labels for electrochemiluminescent detection The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium calibrators and reagents required to perform up to 25 tests in duplicate The assay has a sensitivity of 0 04 pg/mL and a broad dynamic range from 0 06 - 1500 pg/mL This kit allows for the rapid and accurate quantification of IL-1 levels in various human sample types